Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195520.1 | NP_001182449.1 | 4 | 436 | ||
XM_011534780.2 | XP_011533082.1 | 4 | 436 | |||
XM_011534784.2 | XP_011533086.1 | 3 | 294 | |||
XM_011534785.2 | XP_011533087.1 | 3 | 299 | |||
XM_017018659.1 | XP_016874148.1 | 3 | 412 | |||
XM_017018660.1 | XP_016874149.1 | 3 | 412 | |||
XM_017018661.1 | XP_016874150.1 | 3 | 654 | |||
XM_017018662.1 | XP_016874151.1 | 3 | 397 | |||
GenBank mRNA | AV653338.1 | - | 1 | 12 | ||
BX282338.1 | - | 4 | 387 | |||
DB344052.1 | - | 3 | 433 | |||
JQ012741.1 | - | 4 | 359 | |||
JQ012742.1 | - | 4 | 491 |