Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024458.1 | 6 | 540 | |||
GenBank mRNA | AF318337.1 | - | 6 | 540 | ||
AK055081.1 | - | 2 | 2338 | |||
AK124332.1 | - | 4 | 455 |