Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193636.1 | NP_001180565.1 | 1 | 136 | ||
NM_001193637.1 | NP_001180566.1 | 1 | 136 | |||
GenBank mRNA | AY616183.1 | - | 1 | 71 | ||
AY920361.1 | - | 1 | 71 | |||
AY920362.1 | - | 1 | 71 | |||
AY943857.1 | - | 1 | 71 | |||
BC041992.1 | - | 1 | 63 | |||
DA738374.1 | - | 1 | 136 | |||
KC237374.1 | - | 1 | 71 | |||
KC237375.1 | - | 1 | 71 | |||
KU141377.1 | - | 1 | 71 |
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277864.1 | NP_001264793.1 | 1 | 130 | ||
NR_102687.1 | 1 | 130 | ||||
NR_102688.1 | 1 | 130 | ||||
GenBank mRNA | GQ871924.1 | - | 1 | 79 | ||
KU058702.1 | - | 1 | 71 | |||
KU058703.1 | - | 1 | 71 | |||
KU058704.1 | - | 1 | 71 | |||
KU058705.1 | - | 1 | 71 | |||
KU058706.1 | - | 1 | 71 | |||
KU058707.1 | - | 1 | 71 | |||
KU058708.1 | - | 1 | 71 | |||
KU058709.1 | - | 1 | 71 |