Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277864.1 | NP_001264793.1 | 1 | 373 | ||
NR_102687.1 | 1 | 373 | ||||
NR_102688.1 | 1 | 373 | ||||
GenBank mRNA | KU058702.1 | - | 1 | 314 | ||
KU058703.1 | - | 1 | 314 | |||
KU058704.1 | - | 1 | 314 | |||
KU058705.1 | - | 1 | 314 | |||
KU058706.1 | - | 1 | 314 | |||
KU058707.1 | - | 1 | 314 | |||
KU058709.1 | - | 1 | 314 |
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY616183.1 | - | 1 | 314 | ||
AY920361.1 | - | 1 | 314 | |||
AY920362.1 | - | 1 | 314 | |||
AY943857.1 | - | 1 | 314 | |||
BC041992.1 | - | 1 | 306 | |||
DA738374.1 | - | 1 | 379 | |||
DN237881.1 | - | 1 | 66 | |||
GU564235.1 | - | 1 | 146 | |||
KC237374.1 | - | 1 | 314 | |||
KC237375.1 | - | 1 | 314 | |||
KU141377.1 | - | 1 | 314 |