Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100288637 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038253.1 | 4, 5 | 443 | |||
NR_038254.1 | 3, 4 | 343 | ||||
GenBank mRNA | AL578899.3 | - | 1, 2 | 155 | ||
BC112921.1 | - | 2, 3 | 120 | |||
BX954797.1 | - | 3, 4 | 344 |
Gene Symbol: OTUD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126002.1 | - | 4, 5 | 341 | ||
AK308630.1 | - | 3, 4 | 287 | |||
BC035668.1 | - | 2, 3 | 162 |