Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038448.1 | 1, 2 | 841 | |||
GenBank mRNA | BC121819.1 | - | 1, 2 | 505 |