Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004333.4 | NP_001004333.2 | 3 | 478 | ||
NR_037715.1 | 4 | 695 | ||||
NR_037716.1 | 3 | 474 | ||||
GenBank mRNA | AA633512.1 | - | 1 | 327 | ||
AA843796.1 | - | 1 | 336 | |||
AK289930.1 | - | 3 | 261 | |||
AM746459.1 | - | 3 | 245 | |||
BC051802.1 | - | 3 | 249 | 4346419 | ||
BC062705.1 | - | 3 | 253 | 4273688 | ||
BC095436.1 | - | 3 | 243 | |||
BM835723.1 | - | 3 | 478 | |||
BQ223829.1 | - | 4 | 464 | |||
CB160051.1 | - | 3 | 416 | |||
CD176288.1 | - | 3 | 280 | |||
KF980888.1 | - | 3 | 319 |
Gene Symbol: RNASEK-C17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037717.1 | 3 | 474 | |||
GenBank mRNA | BC040148.1 | - | 3 | 231 | 5588971 |