Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC79999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291904.1 | NP_001278833.1 | 3 | 448 | ||
XM_017025119.1 | XP_016880608.1 | 4 | 624 | |||
GenBank mRNA | BU595835.1 | - | 3 | 459 |
Gene Symbol: GRAPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017024635.1 | XP_016880124.1 | 5 | 2649 | ||
XM_017024636.1 | XP_016880125.1 | 4 | 2628 | |||
GenBank mRNA | AK130829.1 | - | 1 | 62 |
Gene Symbol: LOC102724624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524126.2 | XP_011522428.1 | 4 | 637 |
Gene Symbol: LOC388436 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291905.1 | NP_001278834.1 | 3 | 448 | ||
GenBank mRNA | BC058012.1 | - | 3 | 460 |