Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257150.3 | XP_005257207.1 | 19 | 6661 | ||
XM_005257151.3 | XP_005257208.1 | 19 | 6658 | |||
XM_005257152.2 | XP_005257209.1 | 19 | 6661 | |||
XM_005257154.3 | XP_005257211.1 | 19 | 6661 | |||
XM_005257155.3 | XP_005257212.1 | 18 | 6472 | |||
XM_005257156.3 | XP_005257213.1 | 18 | 6472 | |||
XM_005257157.3 | XP_005257214.1 | 18 | 6469 | |||
XM_005257158.3 | XP_005257215.1 | 17 | 6283 | |||
XM_005257159.2 | XP_005257216.1 | 19 | 6661 | |||
XM_005257161.3 | XP_005257218.1 | 19 | 6661 | |||
XM_011524520.2 | XP_011522822.1 | 19 | 6658 | |||
XM_011524521.2 | XP_011522823.1 | 18 | 6469 | |||
XM_011524522.2 | XP_011522824.1 | 17 | 6280 | |||
XM_011524524.2 | XP_011522826.1 | 11 | 3622 | |||
XM_017024353.1 | XP_016879842.1 | 19 | 6661 | |||
XM_017024354.1 | XP_016879843.1 | 13 | 5390 |