Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257970.3 | XP_005258027.1 | 20 | 6055 | ||
GenBank mRNA | AK091492.1 | - | 1 | 1127 |