Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257150.3 | XP_005257207.1 | 28 | 9101 | ||
XM_005257151.3 | XP_005257208.1 | 28 | 9098 | |||
XM_005257152.2 | XP_005257209.1 | 28 | 9101 | |||
XM_005257153.3 | XP_005257210.1 | 27 | 8939 | |||
XM_005257155.3 | XP_005257212.1 | 27 | 8912 | |||
XM_005257156.3 | XP_005257213.1 | 27 | 8912 | |||
XM_005257157.3 | XP_005257214.1 | 27 | 8909 | |||
XM_005257158.3 | XP_005257215.1 | 26 | 8723 | |||
XM_005257159.2 | XP_005257216.1 | 28 | 8672 | |||
XM_005257160.2 | XP_005257217.1 | 27 | 8510 | |||
XM_011524520.2 | XP_011522822.1 | 28 | 9098 | |||
XM_011524521.2 | XP_011522823.1 | 27 | 8909 | |||
XM_011524522.2 | XP_011522824.1 | 26 | 8720 | |||
XM_011524523.2 | XP_011522825.1 | 25 | 8558 | |||
XM_011524524.2 | XP_011522826.1 | 20 | 6062 | |||
XM_017024354.1 | XP_016879843.1 | 22 | 7830 | |||
GenBank mRNA | AL833375.1 | - | 6 | 1379 | ||
BC050566.1 | - | 2 | 141 |
Gene Symbol: ARL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC000924.3 | - | 4 | 603 |
Gene Symbol: ARL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011524163.2 | XP_011522465.1 | 7 | 1072 |