Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005256918.4 | XP_005256975.1 | 5 | 1458 | ||
XM_011524162.2 | XP_011522464.1 | 6 | 1721 |