Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135580.1 | NP_001129052.1 | 3 | 438 | ||
XM_011527596.2 | XP_011525898.1 | 3 | 364 | |||
GenBank mRNA | AI138384.1 | - | 2 | 232 | ||
BX396051.2 | - | 3 | 442 | |||
DY654838.1 | - | 2 | 81 |