Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135768.2 | NP_001129240.1 | 8 | 5635 | ||
NM_001135769.2 | NP_001129241.1 | 7 | 5611 | |||
NM_006505.4 | NP_006496.4 | 8 | 5770 | |||
GenBank mRNA | AI374885.1 | - | 1 | 130 | ||
AK098812.1 | - | 1 | 1897 |