Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC728323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125248.1 | - | 7 | 1394 |
Gene Symbol: PSMD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832109.1 | - | 1 | 1747 |