Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017418.2 | NP_001017418.1 | 2 | 272 | ||
XM_017002174.1 | XP_016857663.1 | 2 | 268 | |||
GenBank mRNA | BF090370.1 | - | 1 | 144 |
Gene Symbol: SPRR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005988.2 | NP_005979.1 | 2 | 270 | ||
GenBank mRNA | BC096108.1 | - | 1 | 243 | 40002232 | |
BC096109.1 | - | 1 | 243 | 40002234 | ||
BC096110.1 | - | 1 | 243 | 40002235 | ||
BC096111.1 | - | 1 | 243 | 40002236 | ||
BC128049.1 | - | 2 | 253 | 40115066 | ||
CA433267.1 | - | 1 | 419 |