Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005988.2 | NP_005979.1 | 2 | 228 | ||
GenBank mRNA | AK311939.1 | - | 2 | 229 | ||
BC096108.1 | - | 1 | 201 | 40002232 | ||
BC096109.1 | - | 1 | 201 | 40002234 | ||
BC096110.1 | - | 1 | 201 | 40002235 | ||
BC096111.1 | - | 1 | 201 | 40002236 | ||
BC128049.1 | - | 2 | 211 | 40115066 | ||
BE710686.1 | - | 447 |
Gene Symbol: SPRR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017418.2 | NP_001017418.1 | 2 | 230 | ||
XM_017002174.1 | XP_016857663.1 | 2 | 226 | |||
GenBank mRNA | BF090370.1 | - | 1 | 102 |
Gene Symbol: SPRR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006945.4 | NP_008876.3 | 2 | 227 | ||
XM_017002175.1 | XP_016857664.1 | 2 | 259 | |||
GenBank mRNA | AK291317.1 | - | 2 | 229 | ||
BC093801.1 | - | 2 | 211 | |||
BC093803.1 | - | 2 | 212 | |||
BC120938.1 | - | 2 | 211 | 40115065 | ||
BG183546.1 | - | 3 | 456 | |||
M21302.1 | - | 2 | 227 |