Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045630.1 | 4 | 1380 | |||
GenBank mRNA | AA805263.1 | - | 1 | 127 | ||
BC032239.1 | - | 4 | 1341 |