Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012474.4 | NP_036606.2 | 7 | 4598 | ||
XM_011509970.2 | XP_011508272.2 | 5 | 4133 | |||
XM_017002236.1 | XP_016857725.1 | 8 | 4803 | |||
GenBank mRNA | BM714864.1 | - | 1 | 150 |