Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206491.1 | NP_001193420.1 | 6 | 5472 | ||
NM_020746.4 | NP_065797.2 | 7 | 5647 | |||
NR_037921.1 | 6 | 5474 | ||||
GenBank mRNA | AK056260.1 | - | 1 | 1349 |