Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161841.1 | NP_001155313.1 | 1 | 42 | ||
NM_198596.2 | NP_940998.2 | 1 | 42 | |||
XM_006723828.1 | XP_006723891.1 | 1 | 55 | |||
XM_006723829.2 | XP_006723892.1 | 1 | 55 | |||
XM_011528916.1 | XP_011527218.1 | 1 | 55 |