Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005264230.3 | XP_005264287.1 | 1 | 1471 | ||
XM_011532729.2 | XP_011531031.1 | 1 | 1471 | |||
XM_011532731.2 | XP_011531033.1 | 1 | 1471 |