Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145054.1 | NP_001138526.1 | 2 | 418 | ||
NM_001316764.1 | NP_001303693.1 | 2 | 170 | |||
NM_001316766.1 | NP_001303695.1 | 2 | 170 | |||
GenBank mRNA | BM045416.1 | - | 2 | 421 | ||
BU184705.1 | - | 2 | 131 | |||
HY050307.1 | - | 2 | 172 |