Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001370.1 | NP_001361.1 | 19 | 3065 | ||
XM_006711956.2 | XP_006712019.1 | 19 | 3170 | |||
XM_011532649.2 | XP_011530951.1 | 19 | 3019 | |||
XM_011532650.2 | XP_011530952.1 | 19 | 3020 | |||
XM_011532652.1 | XP_011530954.1 | 19 | 3065 | |||
XM_011532653.1 | XP_011530955.1 | 19 | 3065 | |||
XM_011532654.1 | XP_011530956.1 | 18 | 2798 | |||
XM_011532655.2 | XP_011530957.1 | 15 | 2087 | |||
XM_011532657.1 | XP_011530959.1 | 13 | 1826 | |||
XM_011532660.1 | XP_011530962.1 | 19 | 3065 | |||
XM_017003521.1 | XP_016859010.1 | 19 | 3065 | |||
XM_017003522.1 | XP_016859011.1 | 14 | 2891 | |||
XM_017003523.1 | XP_016859012.1 | 14 | 2058 | |||
XM_017003524.1 | XP_016859013.1 | 19 | 3065 |