Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128929.3 | NP_001122401.1 | 1 | 196 | ||
XM_017006990.1 | XP_016862479.1 | 1 | 196 | |||
XM_017006992.1 | XP_016862481.1 | 1 | 196 | |||
XM_017006996.1 | XP_016862485.1 | 1 | 196 | |||
XM_017006997.1 | XP_016862486.1 | 1 | 196 | |||
XM_017007004.1 | XP_016862493.1 | 1 | 196 | |||
XM_017007005.1 | XP_016862494.1 | 1 | 196 | |||
GenBank mRNA | AK301176.1 | - | 1 | 69 | ||
DQ533873.1 | - | 1 | 177 |