Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017554.2 | NP_060024.2 | 1 | 218 | ||
XM_011512928.2 | XP_011511230.1 | 1 | 309 | |||
XM_011512929.2 | XP_011511231.1 | 1 | 317 | |||
GenBank mRNA | AK304269.1 | - | 1 | 186 | ||
CR977108.1 | - | 1 | 178 | |||
DC320673.1 | - | 1 | 220 |