Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034845.2 | NP_001030017.2 | 2 | 582 | ||
XM_017008243.1 | XP_016863732.1 | 2 | 1316 | |||
GenBank mRNA | AK124240.1 | - | 2 | 1752 | ||
BC047551.1 | - | 2 | 503 | 5301536 |