Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114357.1 | NP_001107829.1 | 2 | 133 | ||
XM_005263019.3 | XP_005263076.1 | 4 | 486 | |||
XM_011531989.2 | XP_011530291.1 | 3 | 412 | |||
XM_017008236.1 | XP_016863725.1 | 4 | 1879 | |||
XM_017008237.1 | XP_016863726.1 | 5 | 654 | |||
XM_017008238.1 | XP_016863727.1 | 3 | 1663 | |||
XM_017008239.1 | XP_016863728.1 | 3 | 1666 | |||
XM_017008240.1 | XP_016863729.1 | 3 | 415 | |||
XM_017008242.1 | XP_016863731.1 | 4 | 1945 | |||
GenBank mRNA | BC141967.1 | - | 6 | 839 | ||
BC150574.1 | - | 2 | 133 | 40127870 |