Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109825.1 | 1 | 317 | |||
GenBank mRNA | BC144330.1 | - | 1 | 317 | ||
BC144331.1 | - | 1 | 317 |