Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2F1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109819.1 | 1 | 682 | |||
NR_109822.1 | 1 | 682 | ||||
GenBank mRNA | AK296281.1 | - | 1 | 369 | ||
AK311323.1 | - | 1 | 395 |