Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001019.4 | NP_001010.2 | 4, 5 | 397 | ||
NM_001030009.1 | NP_001025180.1 | 4, 5 | 331 | |||
GenBank mRNA | AB062400.1 | - | 1 | 313 | ||
AK130967.1 | - | 4, 5 | 331 | |||
AK291988.1 | - | 4, 5 | 397 | |||
AK311976.1 | - | 1 | 331 | |||
AY208299.1 | - | 3, 4 | 307 | |||
BC001697.2 | - | 4, 5 | 366 | 2967511 | ||
BC030569.1 | - | 3, 4 | 329 | 5580542 | ||
BC046113.1 | - | 1 | 314 | 5736078 | ||
BC105273.1 | - | 1 | 337 | 3839617 | ||
BC105292.1 | - | 4, 5 | 366 | 6649512 | ||
BF678907.1 | - | 4, 5 | 398 | |||
BG035607.1 | - | 384 | ||||
X84407.1 | - | 1 | 301 |