Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGC72080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002822.3 | 4 | 528 | |||
GenBank mRNA | AA574056.1 | - | 1 | 192 | ||
BC029615.1 | - | 4 | 538 | |||
BC042035.1 | - | 3 | 467 |
Gene Symbol: DAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HQ179936.1 | - | 4 | 415 | ||
HQ179937.1 | - | 5 | 507 |
Gene Symbol: OR7E12P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_044999.1 | 1 | 430 |
Gene Symbol: OR7E35P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF073924.1 | - | 1 | 84 |
Gene Symbol: OR7E37P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002163.3 | 1 | 417 |