Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100130698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136612.1 | 6 | 1084 | |||
GenBank mRNA | AK125824.1 | - | 1 | 3315 | ||
AK128140.1 | - | 1 | 1800 |
Gene Symbol: LOC729815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136624.1 | 3 | 1473 | |||
GenBank mRNA | AK302451.1 | - | 3 | 1413 |