Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146162.1 | NP_001139634.1 | 5, 6 | 867 | ||
NM_001271942.1 | NP_001258871.1 | 4, 5 | 636 | |||
GenBank mRNA | JF968445.1 | - | 4, 5 | 414 | ||
JF968446.1 | - | 4, 5 | 285 |