Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195223.1 | NP_001182152.1 | 2 | 166 | ||
XM_017005844.1 | XP_016861333.1 | 2 | 3430 | |||
XM_017005845.1 | XP_016861334.1 | 2 | 56 | |||
GenBank mRNA | BI085096.1 | - | 2 | 158 |