Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005266485.2 | XP_005266542.1 | 1 | 50 | ||
XM_011535177.2 | XP_011533479.1 | 1 | 60 | |||
XM_011535178.1 | XP_011533480.1 | 1 | 50 | |||
XM_017020685.1 | XP_016876174.1 | 1 | 50 |