Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027992.1 | 1 | 73 | |||
GenBank mRNA | DA278599.1 | - | 1 | 73 |
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021090.3 | NP_066576.1 | 1 | 42 | ||
NM_153050.2 | NP_694690.1 | 1 | 42 | |||
NM_153051.2 | NP_694691.1 | 1 | 42 | |||
XM_005261803.2 | XP_005261860.1 | 1 | 42 | |||
XM_005261804.3 | XP_005261861.1 | 1 | 46 | |||
XM_005261805.2 | XP_005261862.1 | 1 | 42 | |||
XM_005261806.2 | XP_005261863.1 | 1 | 42 | |||
XM_005261808.2 | XP_005261865.1 | 1 | 42 | |||
XM_017029034.1 | XP_016884523.1 | 1 | 15 | |||
GenBank mRNA | AK289581.1 | - | 1 | 42 | ||
BC151217.1 | - | 1 | 56 | 40148098 | ||
DA126318.1 | - | 1 | 42 |