Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL832622.2 | - | 1 | 93 | ||
BI552657.1 | - | 1 | 76 | |||
DB062330.1 | - | 1 | 71 |