Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FER1L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293083.1 | NP_001280012.1 | 7 | 652 | ||
XM_011512110.2 | XP_011510412.1 | 7 | 669 | |||
XM_011512111.2 | XP_011510413.1 | 7 | 669 | |||
XM_011512113.2 | XP_011510415.1 | 7 | 669 | |||
XM_011512114.2 | XP_011510416.1 | 7 | 669 | |||
XM_011512115.2 | XP_011510417.1 | 7 | 669 | |||
XM_011512116.2 | XP_011510418.1 | 7 | 669 | |||
XM_011512117.2 | XP_011510419.1 | 7 | 669 | |||
XM_011512118.2 | XP_011510420.1 | 7 | 669 | |||
XM_011512119.2 | XP_011510421.1 | 7 | 669 | |||
XM_011512120.2 | XP_011510422.1 | 6 | 560 | |||
XM_011512122.2 | XP_011510424.1 | 7 | 670 | |||
XM_011512125.2 | XP_011510427.1 | 7 | 670 | |||
XM_011512126.2 | XP_011510428.1 | 7 | 669 | |||
XM_011512127.2 | XP_011510429.1 | 7 | 670 | |||
XM_011512128.2 | XP_011510430.1 | 7 | 669 | |||
XM_017005222.1 | XP_016860711.1 | 7 | 669 | |||
GenBank mRNA | AK126032.1 | - | 6 | 560 | ||
AY461813.1 | - | 7 | 552 |