Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034845.2 | NP_001030017.2 | 10, 11 | 2113 | ||
XM_011531993.2 | XP_011530295.1 | 9, 10 | 1451 | |||
XM_011531994.1 | XP_011530296.1 | 9, 10 | 1285 | |||
XM_011531995.1 | XP_011530297.1 | 8, 9 | 1196 | |||
XM_011531996.1 | XP_011530298.1 | 7, 8 | 1096 | |||
XM_011531997.1 | XP_011530299.1 | 7, 8 | 1018 | |||
XM_017008244.1 | XP_016863733.1 | 9, 10 | 2219 | |||
GenBank mRNA | AJ626725.1 | - | 9, 10 | 1326 | ||
BC047551.1 | - | 10, 11 | 2034 | 5301536 | ||
GU220060.1 | - | 9, 10 | 1377 |