Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105375355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250612.3 | XP_005250669.1 | 4 | 2412 | ||
XM_006716140.3 | XP_006716203.1 | 6 | 2084 | |||
XM_006716141.3 | XP_006716204.1 | 5 | 2004 |