Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 836 | |||
NR_110066.1 | 3 | 518 | ||||
NR_110067.1 | 4 | 774 | ||||
NR_110068.1 | 2 | 474 | ||||
NR_110069.1 | 3 | 666 | ||||
NR_110070.1 | 5 | 625 | ||||
GenBank mRNA | AK090593.1 | - | 3 | 400 | ||
BM544796.1 | - | 5 | 646 | |||
DA025114.1 | - | 3 | 400 | |||
DA689880.1 | - | 2 | 354 |