Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT7P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024271.1 | 1 | 305 | |||
GenBank mRNA | AL832059.1 | - | 1 | 241 | ||
BC092503.1 | - | 1 | 250 | |||
DA100871.1 | - | 1 | 305 |
Gene Symbol: LOC100287497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136590.1 | 1 | 264 | |||
NR_136591.1 | 1 | 264 | ||||
NR_136592.1 | 1 | 264 | ||||
NR_136593.1 | 1 | 264 | ||||
GenBank mRNA | DA132394.1 | - | 1 | 247 | ||
DB083170.1 | - | 1 | 265 |