Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008777.2 | NP_001008777.2 | 7 | 941 | ||
XM_011524865.2 | XP_011523167.1 | 7 | 936 | |||
XM_011524866.2 | XP_011523168.1 | 6 | 843 | |||
XM_011524867.2 | XP_011523169.1 | 7 | 1014 | |||
GenBank mRNA | AY700575.1 | - | 8 | 941 | ||
BC140879.1 | - | 8 | 911 | 9021747 | ||
BC140880.1 | - | 8 | 911 | 9021748 |