Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014451.2 | NP_001014451.1 | 1, 2 | 197 | ||
NM_007242.5 | NP_009173.1 | 1, 2 | 197 | |||
XM_011522832.2 | XP_011521134.1 | 2, 3 | 253 | |||
XM_011522833.2 | XP_011521135.1 | 2, 3 | 253 | |||
GenBank mRNA | AJ237946.1 | - | 1, 2 | 58 | ||
AK027378.1 | - | 1, 2 | 162 | |||
AK223115.1 | - | 1, 2 | 168 | |||
AL136639.1 | - | 1, 2 | 166 | |||
BC003626.2 | - | 1, 2 | 161 | 3030038 | ||
CR457215.1 | - | 1, 2 | 58 | |||
CR533509.1 | - | 1, 2 | 58 | |||
DC385044.1 | - | 1, 2 | 200 |