Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017437.3 | NP_001017437.2 | 5 | 1677 | ||
NM_001318334.1 | NP_001305263.1 | 5 | 1723 | |||
XM_006724278.3 | XP_006724341.1 | 5 | 1284 | |||
XM_006724279.3 | XP_006724342.1 | 5 | 1284 | |||
XM_006724280.3 | XP_006724343.1 | 5 | 1284 | |||
XM_011530263.2 | XP_011528565.1 | 5 | 1337 | |||
XM_011530266.2 | XP_011528568.1 | 5 | 1284 | |||
XM_011530267.2 | XP_011528569.1 | 5 | 1284 | |||
XM_011530268.2 | XP_011528570.1 | 5 | 1284 | |||
XM_011530270.2 | XP_011528572.1 | 5 | 1284 | |||
GenBank mRNA | BC092419.1 | - | 5 | 1311 | 30383661 | |
CR936833.1 | - | 4 | 939 |
Gene Symbol: KIAA1656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046312.1 | 2 | 5493 | |||
GenBank mRNA | AB051443.1 | - | 1 | 4858 |