Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012392.3 | NP_036524.1 | 2 | 471 | ||
NR_033688.1 | 2 | 312 | ||||
XM_011541745.1 | XP_011540047.1 | 2 | 98 | |||
XM_011541746.2 | XP_011540048.1 | 2 | 565 | |||
XM_011541747.1 | XP_011540049.1 | 2 | 440 | |||
XM_017001680.1 | XP_016857169.1 | 2 | 429 | |||
XM_017001681.1 | XP_016857170.1 | 2 | 107 | |||
GenBank mRNA | AB018357.1 | - | 2 | 110 | ||
AB026628.1 | - | 2 | 110 | |||
AK001420.1 | - | 2 | 123 | |||
AK301226.1 | - | 2 | 107 | |||
AY359011.1 | - | 2 | 114 | |||
BC002773.2 | - | 2 | 100 | 3615612 | ||
BC012561.1 | - | 2 | 116 | 4273213 | ||
BI826532.1 | - | 2 | 284 | |||
CR542139.1 | - | 2 | 98 | |||
DB024828.1 | - | 2 | 313 | |||
DC301714.1 | - | 2 | 471 |