Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127688.1 | - | 1 | 659 | ||
BC132990.1 | - | 1 | 37 | |||
BC132992.1 | - | 1 | 37 | 40147014 |