Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031672.2 | NP_001026842.2 | 3 | 391 | ||
GenBank mRNA | AK090456.1 | - | 3 | 329 | ||
BC012781.2 | - | 3 | 324 | |||
BC049198.1 | - | 1 | 48 | |||
BC064419.1 | - | 3 | 354 | |||
BC071735.1 | - | 3 | 330 | |||
CD515193.1 | - | 1 | 137 | |||
DA793739.1 | - | 3 | 391 |