Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033018.2 | NP_001028190.2 | 1 | 16 | ||
GenBank mRNA | AY621333.1 | - | 1 | 16 | ||
DQ012026.1 | - | 1 | 16 |